Assay Details
| Gene Symbol: Tox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145711.4 | NP_663757.3 | 7 - 8 | 1624 | 94 | |
| GenBank mRNA | AF472514.1 | - | 7 - 8 | 1420 | 94 | |
| AK029694.1 | - | 7 - 8 | 1421 | 94 | ||
| AK038671.1 | - | 7 - 8 | 1627 | 94 | ||
| AK133397.1 | - | 7 - 8 | 1421 | 94 | ||
| AK161578.1 | - | 7 - 8 | 1410 | 94 | ||
| BC038873.1 | - | 3 - 4 | 640 | 94 | ||
| BC080732.1 | - | 7 - 8 | 1541 | 6518356 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tox
Genomic Map