Assay Details
| Gene Symbol: Fbxl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159963.1 | NP_001153435.1 | 9 - 10 | 1947 | 70 | |
| GenBank mRNA | AK085100.1 | - | 9 - 10 | 1920 | 70 | |
| AK088067.1 | - | 9 - 10 | 1914 | 70 | ||
| AK150664.1 | - | 9 - 10 | 1904 | 70 | ||
| AK159744.1 | - | 9 - 10 | 1936 | 70 | ||
| BC023291.1 | - | 7 - 8 | 1552 | 70 | ||
| BC047214.1 | - | 1860 | 6466511 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fbxl5
Genomic Map