Assay Details
| Gene Symbol: Bst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009763.3 | NP_033893.2 | 1 - 2 | 201 | 61 | |
| GenBank mRNA | AK041059.1 | - | 1 - 2 | 180 | 61 | |
| BC033422.1 | - | 1 - 2 | 176 | 61 | ||
| D31788.1 | - | 1 - 2 | 180 | 61 | ||
| L32812.1 | - | 1 - 2 | 184 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bst1
Genomic Map