Assay Details
| Gene Symbol: Atp11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293667.1 | NP_001280596.1 | 12 - 13 | 1522 | 73 | |
| NM_001293668.1 | NP_001280597.1 | 12 - 13 | 1522 | 73 | ||
| NM_015804.3 | NP_056619.1 | 12 - 13 | 1522 | 73 | ||
| NR_121603.1 | 12 - 13 | 1522 | 73 | |||
| GenBank mRNA | AF156551.1 | - | 12 - 13 | 1314 | 73 | |
| AK147497.1 | - | 12 - 13 | 1525 | 73 | ||
| AK173088.1 | - | 12 - 13 | 1675 | 73 | ||
| BC138715.1 | - | 12 - 13 | 1241 | 8861737 | 73 | |
| BC145399.1 | - | 12 - 13 | 1242 | 73 | ||
| BC145400.1 | - | 12 - 13 | 1242 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp11a
Genomic Map