Assay Details
| Gene Symbol: Tubgcp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198031.1 | NP_932148.1 | 14 - 15 | 1813 | 85 | |
| GenBank mRNA | AK150149.1 | - | 14 - 15 | 1837 | 85 | |
| AK171491.1 | - | 14 - 15 | 1819 | 85 | ||
| BC025647.1 | - | 9 - 10 | 1060 | 85 | ||
| BC046615.1 | - | 13 - 14 | 1680 | 85 | ||
| BC058566.1 | - | 14 - 15 | 1813 | 6466057 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tubgcp3
Genomic Map