Assay Details
| Gene Symbol: 4930448K20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_004448.2 | 1 | 1208 | 91 | ||
| GenBank mRNA | AK015422.1 | - | 1 | 1211 | 91 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930448K20Rik
Genomic Map