Assay Details
| Gene Symbol: Pdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033453.3 | NP_001028625.1 | 1 - 2 | 150 | 117 | |
| GenBank mRNA | AK133542.1 | - | 1 - 2 | 94 | 117 | |
| AK137550.1 | - | 1 - 2 | 123 | 117 | ||
| BY101287.1 | - | 1 - 2 | 154 | 117 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK137550.1, NM_001033453.3 transcripts.
Set Membership: |
|

Pdp1
Genomic Map