Assay Details
| Gene Symbol: Chat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009891.2 | NP_034021.1 | 12 - 13 | 1641 | 97 | |
| GenBank mRNA | AK046406.1 | - | 12 - 13 | 1642 | 97 | |
| AK047708.1 | - | 11 - 12 | 1533 | 97 | ||
| BC119322.1 | - | 11 - 12 | 1403 | 8734071 | 97 | |
| BC120589.1 | - | 11 - 12 | 1403 | 40129512 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chat
Genomic Map