Assay Details
| Gene Symbol: Chat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009891.2 | NP_034021.1 | 3 - 4 | 390 | 67 | |
| GenBank mRNA | AK046406.1 | - | 3 - 4 | 391 | 67 | |
| AK047708.1 | - | 2 - 3 | 282 | 67 | ||
| AK143147.1 | - | 2 - 3 | 288 | 67 | ||
| BC119322.1 | - | 2 - 3 | 152 | 8734071 | 67 | |
| BC120589.1 | - | 2 - 3 | 152 | 40129512 | 67 | |
| D12487.1 | - | 2 - 3 | 314 | 67 | ||
| D12488.1 | - | 3 - 4 | 388 | 67 | ||
| D12489.1 | - | 2 - 3 | 199 | 67 | ||
| D12490.1 | - | 2 - 3 | 283 | 67 | ||
| D12491.1 | - | 2 - 3 | 170 | 67 | ||
| D12492.1 | - | 3 - 4 | 438 | 67 | ||
| D12493.1 | - | 3 - 4 | 359 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chat
Genomic Map