Assay Details
| Gene Symbol: Ogdhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081130.1 | NP_001074599.1 | 15 - 16 | 2085 | 56 | |
| GenBank mRNA | BC157970.1 | - | 14 - 15 | 1885 | 9007292 | 56 |
| BC157971.1 | - | 14 - 15 | 1885 | 9007293 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163354 | BC157970.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163354 | BC157971.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163354 | NM_001081130.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163354 | BC157970.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163354 | BC157971.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P163354 | NM_001081130.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389862 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 132710 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510370 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 114329 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 753869 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811182 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 501242 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904236 | AF121978.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303940 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411770 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19990 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499587 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 744990 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423942 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190391 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 124546 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119891 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163354 | BC157970.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163354 | BC157971.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163354 | NM_001081130.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163354 | BC157970.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163354 | BC157971.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P163354 | NM_001081130.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163354 | BC157970.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163354 | BC157971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163354 | NM_001081130.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163354 | BC157970.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163354 | BC157971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P163354 | NM_001081130.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29516981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11162411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29517009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29517007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29517022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11735983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29517011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29517010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29517027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11162413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29517000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29517008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11393074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11162409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11770912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11245215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11279384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29517004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11133001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11393070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29517005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11704752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29517021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11704755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29517023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29517017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11245210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11245204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11245212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11336543 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11245221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29517015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29516963 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10413881 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413883 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413885 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413889 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413893 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413888 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413887 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413895 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413880 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413878 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413882 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413875 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413879 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413884 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413877 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413894 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413896 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413876 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413890 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413891 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413886 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413892 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10413874 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400003045.mm.1 | NM_001081130.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400003045.mm.1 | NM_001081130.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400003045.mm.1 | TC1400000344.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 614584 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2924146 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2924146 | - | - |

Ogdhl
Genomic Map