Assay Details
| Gene Symbol: Ogdhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081130.1 | NP_001074599.1 | 4 - 5 | 545 | 60 | |
| GenBank mRNA | BC157970.1 | - | 3 - 4 | 402 | 9007292 | 60 |
| BC157971.1 | - | 3 - 4 | 402 | 9007293 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ogdhl
Genomic Map