Assay Details
| Gene Symbol: Ogdhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081130.1 | NP_001074599.1 | 7 - 8 | 978 | 86 | |
| GenBank mRNA | BC157970.1 | - | 6 - 7 | 778 | 9007292 | 86 |
| BC157971.1 | - | 6 - 7 | 778 | 9007293 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ogdhl
Genomic Map