Assay Details
| Gene Symbol: Gab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181584.4 | NP_853615.1 | 7 - 8 | 1584 | 108 | |
| GenBank mRNA | AJ748654.1 | - | 4 - 5 | 431 | 108 | |
| AK031227.1 | - | 7 - 8 | 1582 | 108 | ||
| AY057988.1 | - | 7 - 8 | 1470 | 108 | ||
| BB634123.1 | - | 4 - 5 | 579 | 108 | ||
| BC141240.1 | - | 7 - 8 | 1464 | 9055852 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gab3
Genomic Map