Assay Details
| Gene Symbol: Gm39127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045563.1 | - | 2 - 3 | 347 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | arginine and glutamate rich 1 | Arglu1 |
9430010O03Rik, C130008N12 |
-58.6kb | Anti-sense | Chr. 8 - 8666567 - 8690553 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4933430N04 gene | 4933430N04Rik | 370.1kb | Anti-sense | Chr. 8 - 9125412 - 9132288 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm39127
Genomic Map