Assay Details
| Gene Symbol: Tbl1x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020601.2 | NP_065626.1 | 10 - 11 | 1161 | 70 | |
| GenBank mRNA | AK030547.1 | - | 10 - 11 | 1376 | 70 | |
| AK031937.1 | - | 6 - 7 | 622 | 70 | ||
| AK038674.1 | - | 10 - 11 | 1162 | 70 | ||
| BC043105.1 | - | 10 - 11 | 1284 | 6406579 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbl1x
Genomic Map