Assay Details
| Gene Symbol: Tbl1x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020601.2 | NP_065626.1 | 6 - 7 | 794 | 80 | |
| GenBank mRNA | AK030547.1 | - | 6 - 7 | 1009 | 80 | |
| AK031937.1 | - | 2 - 3 | 255 | 80 | ||
| AK036996.1 | - | 7 - 8 | 873 | 80 | ||
| AK038674.1 | - | 6 - 7 | 795 | 80 | ||
| BC043105.1 | - | 6 - 7 | 917 | 6406579 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbl1x
Genomic Map