Assay Details
| Gene Symbol: Tbl1x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020601.2 | NP_065626.1 | 7 - 8 | 931 | 96 | |
| GenBank mRNA | AK030547.1 | - | 7 - 8 | 1146 | 96 | |
| AK031937.1 | - | 3 - 4 | 392 | 96 | ||
| AK038674.1 | - | 7 - 8 | 932 | 96 | ||
| BC043105.1 | - | 7 - 8 | 1054 | 6406579 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbl1x
Genomic Map