Assay Details
| Gene Symbol: Grid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008166.2 | NP_032192.2 | 10 - 11 | 1625 | 70 | |
| GenBank mRNA | AK138279.1 | - | 10 - 11 | 1615 | 70 | |
| D10171.1 | - | 10 - 11 | 1595 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grid1
Genomic Map