Assay Details
| Gene Symbol: Grid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008166.2 | NP_032192.2 | 8 - 9 | 1323 | 84 | |
| GenBank mRNA | AK138279.1 | - | 8 - 9 | 1313 | 84 | |
| D10171.1 | - | 8 - 9 | 1293 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grid1
Genomic Map