Assay Details
| Gene Symbol: Slc20a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011394.3 | NP_035524.2 | 4 - 5 | 1009 | 134 | |
| GenBank mRNA | AF196476.1 | - | 3 - 4 | 777 | 134 | |
| AK087507.1 | - | 4 - 5 | 834 | 134 | ||
| AK157206.1 | - | 4 - 5 | 883 | 134 | ||
| AK170985.1 | - | 4 - 5 | 975 | 134 | ||
| BC046510.1 | - | 4 - 5 | 932 | 4035182 | 134 | |
| BC094562.1 | - | 4 - 5 | 1389 | 134 | ||
| U62559.1 | - | 3 - 4 | 488 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc20a2
Genomic Map