Assay Details
| Gene Symbol: Slc20a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011394.3 | NP_035524.2 | 6 - 7 | 1219 | 62 | |
| GenBank mRNA | AF196476.1 | - | 5 - 6 | 987 | 62 | |
| AK087507.1 | - | 6 - 7 | 1044 | 62 | ||
| AK157206.1 | - | 6 - 7 | 1093 | 62 | ||
| AK170985.1 | - | 6 - 7 | 1185 | 62 | ||
| AK198610.1 | - | 4 - 5 | 383 | 62 | ||
| AK216704.1 | - | 2 - 3 | 230 | 62 | ||
| AK220047.1 | - | 3 - 4 | 248 | 62 | ||
| BC046510.1 | - | 6 - 7 | 1142 | 4035182 | 62 | |
| BC094562.1 | - | 6 - 7 | 1599 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc20a2
Genomic Map