Assay Details
| Gene Symbol: Ikbkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159774.1 | NP_001153246.1 | 10 - 11 | 1060 | 64 | |
| NM_010546.2 | NP_034676.1 | 10 - 11 | 1060 | 64 | ||
| GenBank mRNA | AF026524.1 | - | 9 - 10 | 928 | 64 | |
| AF088910.1 | - | 10 - 11 | 1120 | 64 | ||
| AK144603.1 | - | 10 - 11 | 1053 | 64 | ||
| AK150884.1 | - | 10 - 11 | 1012 | 64 | ||
| AK151888.1 | - | 10 - 11 | 1012 | 64 | ||
| AK151955.1 | - | 10 - 11 | 1012 | 64 | ||
| AK153151.1 | - | 10 - 11 | 1012 | 64 | ||
| AK155587.1 | - | 10 - 11 | 993 | 64 | ||
| AK156294.1 | - | 10 - 11 | 1018 | 64 | ||
| BC004772.1 | - | 10 - 11 | 998 | 64 | ||
| BC037723.1 | - | 10 - 11 | 1032 | 4482634 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ikbkb
Genomic Map