| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800014823.mm.1 |
NM_001159774.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014823.mm.1 |
NM_001159774.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014823.mm.1 |
NM_010546.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014823.mm.1 |
NM_010546.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014823.mm.1 |
TC0800001874.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800014820.mm.1 |
NM_001159774.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014820.mm.1 |
NM_001159774.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014820.mm.1 |
NM_010546.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014820.mm.1 |
NM_010546.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014820.mm.1 |
TC0800001874.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800014822.mm.1 |
NM_001159774.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014822.mm.1 |
NM_001159774.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014822.mm.1 |
NM_010546.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014822.mm.1 |
NM_010546.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014822.mm.1 |
TC0800001874.mm.1 |
MTA TC |