Assay Details
| Gene Symbol: Smim19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012667.2 | NP_001012685.2 | 4 - 5 | 694 | 75 | |
| NM_001146117.1 | NP_001139589.1 | 3 - 4 | 635 | 75 | ||
| GenBank mRNA | AK131902.1 | - | 3 - 4 | 386 | 75 | |
| AK133712.1 | - | 3 - 4 | 639 | 75 | ||
| AK140957.1 | - | 3 - 4 | 425 | 75 | ||
| AK213440.1 | - | 2 - 3 | 230 | 75 | ||
| BB618223.1 | - | 3 - 4 | 635 | 75 | ||
| BC048089.1 | - | 4 - 5 | 670 | 6466443 | 75 | |
| BQ922100.1 | - | 4 - 5 | 668 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25711066 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25711064 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25711063 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P95740 | BC048089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P95740 | NM_001012667.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P95740 | NM_001146117.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P95740 | AK131902.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P95740 | AK133712.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P95740 | BC048089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P95740 | NM_001012667.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P95740 | NM_001146117.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P95740 | AK131902.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P95740 | AK133712.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412020 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136739 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96064 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955077 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 113625 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381133 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 314314 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155262 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29636 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123219 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113070 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160675 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P95740 | BC048089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P95740 | NM_001012667.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P95740 | NM_001146117.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P95740 | AK131902.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P95740 | AK133712.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P95740 | BC048089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P95740 | NM_001012667.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P95740 | NM_001146117.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P95740 | AK131902.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P95740 | AK133712.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25711073 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113070 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160675 | - | - |
| Whole Mouse Genome G4122A | A_52_P95740 | BC048089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P95740 | NM_001012667.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P95740 | NM_001146117.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P95740 | AK131902.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P95740 | AK133712.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P95740 | BC048089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P95740 | NM_001012667.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P95740 | NM_001146117.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P95740 | AK131902.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P95740 | AK133712.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105131 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113070 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160675 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06908299 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06908293 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06908296 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06908298 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25711063 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25711066 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25711064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25710999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25711132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06714015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07428663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07223424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25710965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07071696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07027160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06968689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06948130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25710956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25710963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06908304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25710953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711077 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07027149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07225688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25710964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25710961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06968691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07027148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25710967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25711071 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10577532 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10577529 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10577533 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10577531 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10577530 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438199_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10577528 | - | - |
| Mouse Genome 430 2.0 Array | 1438199_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438199_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 102032_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 102032_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800014757.mm.1 | NM_001012667.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800014757.mm.1 | NM_001012667.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800014757.mm.1 | NM_001146117.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800014757.mm.1 | NM_001146117.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800014757.mm.1 | TC0800001870.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438199_PM_at | - | - |
| Mouse Expression Set 430 | 1438199_at | - | - |
| Murine Genome U74v2 Set | 166547_s_at | - | - |
| Murine Genome U74v2 Set | 166547_PM_s_at | - | - |
| Murine Genome U74v2 Set | 166546_at | - | - |
| Murine Genome U74v2 Set | 166546_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438199_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438199_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438199_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438199_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 325294 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106450154 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160963 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3160963 | - | - |
| mouse-6 v1.1 (Illumina) | GI_60592995-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3160963 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160963 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | GI_60592995-S | - | - |

Smim19
Genomic Map