Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800001964.mm.1 |
NM_001081662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001964.mm.1 |
NM_001081662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001964.mm.1 |
TC0800000266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001966.mm.1 |
NM_001081662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001966.mm.1 |
NM_001081662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001966.mm.1 |
TC0800000266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001965.mm.1 |
NM_001081662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001965.mm.1 |
NM_001081662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001965.mm.1 |
TC0800000266.mm.1 |
MTA TC |