Assay Details
Gene Symbol: C86695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081662.1 | NP_001075131.1 | 3 - 4 | 501 | 83 | |
GenBank mRNA | DV648735.1 | - | 3 - 4 | 401 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P13364 | NM_001081662.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P13364 | AK135805.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 258578 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 127413 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 464696 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P13364 | NM_001081662.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P13364 | AK135805.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01689500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25708465 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06886682 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708479 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708422 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708418 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708465 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708442 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07002377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708405 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07263201 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708417 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708402 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07292092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07218699 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07124948 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708476 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708458 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708438 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06972928 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708400 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708408 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708414 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708451 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708445 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708431 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708441 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708467 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708448 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07292103 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708469 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708461 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07292107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07292102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708424 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708426 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708427 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07263199 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708403 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708454 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708420 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708416 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708443 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708435 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708425 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708463 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25708411 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10608702 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1443478_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10608702 | - | - |
Mouse Genome 430 2.0 Array | 1443478_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1443478_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 97476_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 97476_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800001967.mm.1 | NM_001081662.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800001967.mm.1 | NM_001081662.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800001967.mm.1 | TC0800000266.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800001968.mm.1 | NM_001081662.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800001968.mm.1 | NM_001081662.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800001968.mm.1 | TC0800000266.mm.1 | MTA TC |
Mouse Expression Set 430 | 1443478_at | - | - |
Mouse Expression Set 430 | 1443478_PM_at | - | - |
Murine Genome U74v2 Set | 129400_at | - | - |
Murine Genome U74v2 Set | 129400_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443478_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443478_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 883142 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_2533182 | - | - |
