Assay Details
| Gene Symbol: Atp7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007511.2 | NP_031537.2 | 10 - 11 | 2598 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp7b| Gene Symbol: Atp7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007511.2 | NP_031537.2 | 10 - 11 | 2598 | 69 | |
Set Membership: |
|