Assay Details
| Gene Symbol: Nek3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF093416.1 | - | 9 - 10 | 927 | 127 | |
| AF099066.1 | - | 10 - 11 | 949 | 127 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Nek3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF093416.1 | - | 9 - 10 | 927 | 127 | |
| AF099066.1 | - | 10 - 11 | 949 | 127 | ||
Set Membership: |
|