Assay Details
| Gene Symbol: Nek3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162947.1 | NP_001156419.1 | 1 - 2 | 173 | 75 | |
| NM_011848.4 | NP_035978.2 | 2 - 3 | 231 | 75 | ||
| GenBank mRNA | AF093416.1 | - | 1 - 2 | 178 | 75 | |
| AF099066.1 | - | 2 - 3 | 200 | 75 | ||
| BC005411.1 | - | 2 - 3 | 208 | 3590866 | 75 | |
| BU058938.1 | - | 2 - 3 | 231 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nek3
Genomic Map