Assay Details
| Gene Symbol: 4930453L07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073360.1 | 2 - 3 | 1280 | 75 | ||
| GenBank mRNA | AK015455.1 | - | 2 - 3 | 1280 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4933430N04 gene | 4933430N04Rik | -14.6kb | Anti-sense | Chr. 8 - 9125412 - 9132288 on NCBI Build GRCm38 | |
| Downstream |
family with sequence similarity 155, member A |
Fam155a |
6430500D16, AW121567, Tmem28 |
57.0kb | Anti-sense | Chr. 8 - 9206010 - 9773363 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930453L07Rik
Genomic Map