| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500021906.mm.1 |
TC0500002447.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021906.mm.1 |
NR_040682.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021906.mm.1 |
NR_040682.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021906.mm.1 |
NR_040683.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021906.mm.1 |
NR_040683.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021909.mm.1 |
TC0500002447.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021909.mm.1 |
NR_040682.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021909.mm.1 |
NR_040682.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021909.mm.1 |
NR_040683.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021909.mm.1 |
NR_040683.1 |
Refseq |