Array Name |
Probe IDs |
Targets |
Target Type |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
717650 |
AK014488.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
717650 |
AK032283.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
717650 |
AK161026.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
717650 |
AK171453.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
717650 |
BC035538.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
717650 |
BC051166.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
717650 |
BC058767.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
717650 |
D26047.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
717650 |
NM_011081.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
126148 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
678151 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
445298 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
132695 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
20671 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05586123 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P514945 |
AK016620.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P514945 |
BC099582.1 |
mouse_transcriptome |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04301483 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04548099 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730673 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730643 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730612 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730615 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730649 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730705 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730599 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730687 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730684 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730680 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730681 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730606 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730594 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730604 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730676 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730675 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730707 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730659 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730647 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730670 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730651 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04800917 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730701 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730702 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04625856 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04242843 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730650 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730693 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730644 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730646 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730602 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730662 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730674 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730663 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730611 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730682 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730613 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730695 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730660 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730597 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730605 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730697 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730653 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730617 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04721041 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730679 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730691 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730709 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730689 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730667 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730699 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23730607 |
- |
- |