Assay Details
| Gene Symbol: Exoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289770.1 | NP_001276699.1 | 2 - 3 | 287 | 90 | |
| NM_001289771.1 | NP_001276700.1 | 2 - 3 | 287 | 90 | ||
| NM_027270.2 | NP_081546.1 | 2 - 3 | 287 | 90 | ||
| GenBank mRNA | AK042620.1 | - | 2 - 3 | 241 | 90 | |
| AK155980.1 | - | 2 - 3 | 241 | 90 | ||
| BC024678.1 | - | 2 - 3 | 184 | 3661758 | 90 | |
| BC064811.1 | - | 2 - 3 | 275 | 5686167 | 90 | |
| BC087543.1 | - | 2 - 3 | 223 | 6334680 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc1
Genomic Map