Assay Details
| Gene Symbol: Exoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289770.1 | NP_001276699.1 | 8 - 9 | 1231 | 67 | |
| NM_001289771.1 | NP_001276700.1 | 8 - 9 | 1231 | 67 | ||
| NM_027270.2 | NP_081546.1 | 9 - 10 | 1252 | 67 | ||
| GenBank mRNA | AK155980.1 | - | 8 - 9 | 1185 | 67 | |
| BC024678.1 | - | 8 - 9 | 1128 | 3661758 | 67 | |
| BC029330.1 | - | 1 - 2 | 80 | 67 | ||
| BC064811.1 | - | 9 - 10 | 1240 | 5686167 | 67 | |
| BC087543.1 | - | 8 - 9 | 1167 | 6334680 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc1
Genomic Map