Assay Details
| Gene Symbol: 4930565D16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040752.1 | 1 - 2 | 199 | 92 | ||
| GenBank mRNA | AK016226.1 | - | 1 - 2 | 200 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tripartite motif-containing 2 | Trim2 |
mKIAA0517, narf |
-72.9kb | Sense | Chr. 3 - 84160439 - 84305832 on NCBI Build GRCm38 |
| Downstream | FH2 domain containing 1 | Fhdc1 |
6330505N24Rik, Gm126 |
62.3kb | Sense | Chr. 3 - 84442191 - 84480439 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930565D16Rik
Genomic Map