Assay Details
| Gene Symbol: 4930565D16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040752.1 | 3 - 4 | 387 | 69 | ||
| GenBank mRNA | AK016226.1 | - | 3 - 4 | 388 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tripartite motif-containing 2 | Trim2 |
mKIAA0517, narf |
-43.5kb | Sense | Chr. 3 - 84160439 - 84305832 on NCBI Build GRCm38 |
| Downstream | FH2 domain containing 1 | Fhdc1 |
6330505N24Rik, Gm126 |
63.9kb | Sense | Chr. 3 - 84442191 - 84480439 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2325172 | AK016226.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 13734 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809086 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78496 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK010612.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK028264.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK030751.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK083821.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK146109.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK149931.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK151109.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK151418.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK153012.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK153249.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK159177.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK159509.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK160889.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK167794.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK169140.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK182564.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK192718.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK193971.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK197407.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK200102.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK201072.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK203072.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK203915.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK205097.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK205890.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK207080.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK207229.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK207824.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK210080.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK211836.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK211883.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK216761.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK218169.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK218259.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK219821.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | AK220148.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | BC043053.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | BC049114.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | NM_009503.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | XM_985571.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901458 | Z14044.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 958037 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 867298 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 51119 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410533 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168263 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 36989 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04970914 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04970949 | - | - |
| Whole Mouse Genome G4122A | A_52_P818604 | AK016226.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04970914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02948338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03029243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02948238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02948246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290428 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02948332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02948323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02422367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02948233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02546906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02546907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03029257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02948328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02948327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03140055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03015253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03029247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22290369 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1432277_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432277_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432277_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300016916.mm.1 | NR_040752.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300016916.mm.1 | NR_040752.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300016916.mm.1 | TC0300002244.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300016917.mm.1 | NR_040752.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300016917.mm.1 | NR_040752.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300016917.mm.1 | TC0300002244.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1432277_at | - | - |
| Mouse Expression Set 430 | 1432277_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432277_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432277_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 759616 | - | - |

4930565D16Rik
Genomic Map