Assay Details
| Gene Symbol: Tbc1d14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113362.1 | NP_001106833.1 | 1 - 2 | 176 | 106 | |
| GenBank mRNA | AK029986.1 | - | 1 - 2 | 177 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001113362.1 transcripts.
Set Membership: |
|

Tbc1d14
Genomic Map