| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500020678.mm.1 |
TC0500002253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020678.mm.1 |
NM_194344.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020678.mm.1 |
NM_194344.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020684.mm.1 |
TC0500002253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020684.mm.1 |
NM_194344.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020684.mm.1 |
NM_194344.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020681.mm.1 |
TC0500002253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020681.mm.1 |
NM_194344.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020681.mm.1 |
NM_194344.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020682.mm.1 |
TC0500002253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020682.mm.1 |
NM_194344.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020682.mm.1 |
NM_194344.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020676.mm.1 |
TC0500002253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020676.mm.1 |
NM_194344.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020676.mm.1 |
NM_194344.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020683.mm.1 |
TC0500002253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020683.mm.1 |
NM_194344.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020683.mm.1 |
NM_194344.2 |
Refseq |