Assay Details
| Gene Symbol: Sh3tc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194344.2 | NP_919325.2 | 5 - 6 | 871 | 124 | |
| GenBank mRNA | AK028482.1 | - | 1 - 2 | 967 | 124 | |
| AK150064.1 | - | 5 - 6 | 649 | 124 | ||
| AK155330.1 | - | 5 - 6 | 632 | 124 | ||
| AK159089.1 | - | 5 - 6 | 650 | 124 | ||
| AK167283.1 | - | 5 - 6 | 572 | 124 | ||
| AK170396.1 | - | 5 - 6 | 651 | 124 | ||
| AK171209.1 | - | 5 - 6 | 628 | 124 | ||
| AK171242.1 | - | 5 - 6 | 628 | 124 | ||
| BC024909.1 | - | 3 - 4 | 321 | 124 | ||
| BC056461.1 | - | 3 - 4 | 215 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3tc1
Genomic Map