Assay Details
| Gene Symbol: Sh3tc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194344.2 | NP_919325.2 | 6 - 7 | 1019 | 60 | |
| GenBank mRNA | AK028482.1 | - | 2 - 3 | 1115 | 60 | |
| AK150064.1 | - | 6 - 7 | 797 | 60 | ||
| AK155330.1 | - | 6 - 7 | 780 | 60 | ||
| AK159089.1 | - | 6 - 7 | 798 | 60 | ||
| AK167283.1 | - | 6 - 7 | 720 | 60 | ||
| AK170383.1 | - | 6 - 7 | 852 | 60 | ||
| AK170396.1 | - | 6 - 7 | 799 | 60 | ||
| AK171209.1 | - | 6 - 7 | 776 | 60 | ||
| AK171242.1 | - | 6 - 7 | 776 | 60 | ||
| BC024909.1 | - | 4 - 5 | 469 | 60 | ||
| BC056461.1 | - | 4 - 5 | 363 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3tc1
Genomic Map