Assay Details
| Gene Symbol: Lmbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020295.3 | NP_064691.2 | 11 - 12 | 1116 | 85 | |
| GenBank mRNA | AF190665.1 | - | 11 - 12 | 918 | 85 | |
| AK030631.1 | - | 12 - 13 | 1922 | 85 | ||
| AK195974.1 | - | 4 - 5 | 306 | 85 | ||
| BC016110.1 | - | 11 - 12 | 875 | 4481088 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmbr1
Genomic Map