Assay Details
| Gene Symbol: Paxip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018878.3 | NP_061366.2 | 14 - 15 | 2784 | 77 | |
| GenBank mRNA | AK144606.1 | - | 14 - 15 | 2646 | 77 | |
| BC066014.1 | - | 14 - 15 | 2573 | 5704566 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Paxip1
Genomic Map