Assay Details
| Gene Symbol: Triqk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173746.3 | NP_776107.2 | 2 - 3 | 249 | 157 | |
| GenBank mRNA | AK048604.1 | - | 2 - 3 | 228 | 157 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK048604.1, NM_173746.3 transcripts.
Set Membership: |
|

Triqk
Genomic Map