Assay Details
| Gene Symbol: En2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010134.3 | NP_034264.1 | 1 - 2 | 1495 | 84 | |
| GenBank mRNA | AK157766.1 | - | 1 - 2 | 1497 | 84 | |
| L12705.1 | - | 1 - 2 | 711 | 84 | ||
More Information
Set Membership: |
|

En2
Genomic Map