Assay Details
| Gene Symbol: Ccdc66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177111.3 | NP_796085.3 | 5 - 6 | 741 | 115 | |
| GenBank mRNA | AK145297.1 | - | 4 - 5 | 754 | 115 | |
| AK158666.1 | - | 3 - 4 | 259 | 115 | ||
| BC070471.1 | - | 5 - 6 | 741 | 30626499 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK158666.1 transcripts.
Set Membership: |
|

Ccdc66
Genomic Map