Assay Details
| Gene Symbol: Mpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ627041.1 | - | 10 - 11 | 1152 | 82 | |
| AK048532.1 | - | 10 - 11 | 1337 | 82 | ||
| AK081829.1 | - | 10 - 11 | 1269 | 82 | ||
| AK152379.1 | - | 10 - 11 | 1251 | 82 | ||
| AK165261.1 | - | 10 - 11 | 1270 | 82 | ||
| BC013444.1 | - | 10 - 11 | 1320 | 4221660 | 82 | |
| BC138314.1 | - | 10 - 11 | 1210 | 8861334 | 82 | |
| BC138315.1 | - | 10 - 11 | 1210 | 8861335 | 82 | |
| U38196.1 | - | 10 - 11 | 1326 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpp1
Genomic Map