Assay Details
| Gene Symbol: Ccnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019937.3 | NP_064321.2 | 3 - 4 | 610 | 99 | |
| GenBank mRNA | AF159159.1 | - | 3 - 4 | 563 | 99 | |
| AF467251.1 | - | 3 - 4 | 604 | 99 | ||
| AK011629.1 | - | 3 - 4 | 567 | 99 | ||
| AK051380.1 | - | 3 - 4 | 567 | 99 | ||
| BC037204.1 | - | 3 - 4 | 548 | 99 | ||
| BC094383.1 | - | 3 - 4 | 546 | 30604933 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccnl1
Genomic Map