Assay Details
| Gene Symbol: Atp6v0d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175406.3 | NP_780615.2 | 5 - 6 | 709 | 77 | |
| GenBank mRNA | AB088358.1 | - | 5 - 6 | 665 | 77 | |
| AK075745.1 | - | 5 - 6 | 749 | 77 | ||
| AK089507.1 | - | 5 - 6 | 634 | 77 | ||
| AK137586.1 | - | 5 - 6 | 690 | 77 | ||
| AK170072.1 | - | 5 - 6 | 683 | 77 | ||
| AK170405.1 | - | 5 - 6 | 682 | 77 | ||
| AK170578.1 | - | 5 - 6 | 653 | 77 | ||
| AK170659.1 | - | 5 - 6 | 658 | 77 | ||
| AY145896.1 | - | 5 - 6 | 709 | 77 | ||
| AY517482.1 | - | 5 - 6 | 701 | 77 | ||
| BC087899.1 | - | 5 - 6 | 643 | 30504725 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v0d2
Genomic Map