Assay Details
| Gene Symbol: Tmprss11d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145561.2 | NP_663536.1 | 6 - 7 | 553 | 94 | |
| GenBank mRNA | AB053953.1 | - | 6 - 7 | 587 | 94 | |
| AF539752.1 | - | 1 - 2 | 126 | 94 | ||
More Information
Set Membership: |
|

Tmprss11d
Genomic Map