Assay Details
| Gene Symbol: Angpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007426.4 | NP_031452.2 | 1 - 2 | 573 | 90 | |
| GenBank mRNA | AF004326.1 | - | 1 - 2 | 499 | 90 | |
| AK019860.1 | - | 1 - 2 | 562 | 90 | ||
| AK048622.1 | - | 1 - 2 | 530 | 90 | ||
| AK143974.1 | - | 1 - 2 | 560 | 90 | ||
| AK156132.1 | - | 1 - 2 | 544 | 90 | ||
| BC027216.1 | - | 1 - 2 | 526 | 3494566 | 90 | |
| BC138309.1 | - | 1 - 2 | 467 | 8861329 | 90 | |
| BC138313.1 | - | 1 - 2 | 467 | 8861333 | 90 | |
| CN539462.1 | - | 1 - 2 | 574 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Angpt2
Genomic Map